Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Starch Agar
Shop All BD Diagnostic Systems Products
Click to view available options
:
Starch Agar; 500g
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Starch Agar |
| Product Type | Agar |
| Formulation | Beef extract 3g/L, Soluble Starch 10g/L, Agar 12g/L |
| For Use With (Application) | Cultivation of microorganisms being tested For Starch hydrolysis |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction