Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Stock Culture Agar
Shop All BD Diagnostic Systems Products
Click to view available options
:
Stock Agar; 500g
Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Content And Storage | <30°C |
Form | Dehydrated |
Media Type | Stock Culture Agar |
Product Type | Agar |
Formulation | Beef heart infusion from 500g 10g/L, Proteose Peptone 10g/L, Gelatin 10g/L, Isoelectric Casein 5g/L, Dextrose 0.5g/L, Disodium Phosphate 4g/L, Sodium Citrate 3g/L, Agar 7.5g/L |
For Use With (Application) | Isolation of Streptococci |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction