Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Tinsdale Base

Catalog No. df0786171
Click to view available options
:
Tinsdale Base; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used with Tinsdale Enrichment Desiccated in isolating and differentiating Corynebacterium diphtheriae

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Form Dehydrated
Media Type Tinsdale Base
Product Type Microbiology Media
Formulation Bovine Serum 333mL, Horse Serum 380mL, L-Cystine 2g/L, Potassium Tellurite 1.4g/L, Sodium Thiosulfate 2.8g/L
For Use With (Application) Isolation of Corynebacterium diphtheriae
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.