Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Universal Preenrichment Broth

Catalog No. df0235178
Click to view available options
:
Universal Preenrichment Broth; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

For simultaneous preenrichment of sub-lethally injured Listeria and Salmonella

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM
Content And Storage Refrigerate
Form Dehydrated
Media Type Universal Preenrichment Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 5g/L, Proteose Peptone 5g/L, Monopotassium Phosphate 15g/L, Disodium Phosphate 7g/L, Sodium Chloride 5g/L, Dextrose 0.5g/L, Magnesium Sulfate 0.25g/L, Ferric Ammonium Citrate 0.1g/L, Sodium Pyruvate 0.2g/L
For Use With (Application) For recovering sublethally injured Salmonella and Listeria from food products
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.