Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Violet Red Bile Agar
Shop All BD Diagnostic Systems Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Description
- Formulation: Yeast extract 3g/L, Peptone 7g/L, No. 3 Bile Salt 1.5g/L, Lactose 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 2mg/L
Order Info
Prices are for units of one each unless otherwise specified
Certifications
BAM, CCAM, COMPF, ISO, SMD
Specifications
Specifications
| Certifications/Compliance | BAM, CCAM, COMPF, ISO, SMD |
| Color | Violet Red |
| Content And Storage | 2°C to 30°C |
| Form | Dehydrated |
| Media Type | Violet Red Bile Agar |
| Product Type | Agar |
| Formulation | Yeast extract 3g/L, Peptone 7g/L, No. 3 Bile Salt 1.5g/L, Lactose 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 2mg/L |
| For Use With (Application) | For enumerating coliform organisms in dairy products |
| Packaging Type | Pail |
| Quantity | 2 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction