Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Yeast Mold Broth

Catalog No. df0711171 Shop All BD Diagnostic Systems Products
Click to view available options
:
Yeast Mold (YM) Broth; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

For cultivation of yeasts, molds and other aciduric microorganisms. BD Difco™ Dehydrated Culture Media: Yeast Mold Broth is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Yeast Mold Broth

Order Info

Prices are for units of one each unless otherwise specified.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Yeast Mold Broth
Product Type Microbiology Media
Formulation Yeast extract 3g/L, Malt extract 3g/L, Peptone 5g/L, Dextrose 10g/L
For Use With (Application) Cultivation of Yeasts, molds and other Aciduric microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.