Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Haemophilus influenzae Serotyping Antisera

Catalog No. p-3201177 Shop All BD Diagnostic Systems Products
Click to view available options
Product Type:
Antiserum
Description:
Antiserum Poly
Antiserum Type a
Antiserum Type b
Antiserum Type c
Antiserum Type d
Antiserum Type e
Antiserum Type f
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Catalog No. DF2237-50-0

Supplier: BD 222371

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Use in slide agglutination tests for serotyping Haemophilus influenzae

  • Lyophilized
  • Rehydrate to 1mL
  • Type: Antisera
  • Shelf Life: 965 days

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate, store at 2°C to 8°C
Description Antiserum Poly
Product Type Antiserum
For Use With (Application) Use in slide agglutination tests for serotyping Haemophilus influenzae
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.