Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human Neural Cell Sorting Kit

Catalog No. bdb562271 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562271 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562271 Supplier BD Supplier No. 562271
Only null left
Add to Cart
Add to Cart

Includes

  • PE Mouse Anti-Human CD24
  • PerCP-Cy5.5 Mouse Anti-Human CD271
  • PerCP-Cy5.5 Mouse Anti-Human CD44
  • PE-Cy7 Mouse Anti-Human CD15
  • APC Mouse Anti-Human CD184
  • PE Mouse IgG1, k Isotype Control
  • PE-Cy7 Mouse IgM, k Isotype Control
  • APC Mouse IgG1, k Isotype Control
  • Negative Control CompBead Plus
  • Anti-Mouse Ig, k CompBead Plus
  • PerCP-Cy5.5 Mouse IgG1, k Isotype Control

Designed to allow the isolation of neural stem cells (NSCs) derived from human pluripotent stem cells or the isolation of neurons and glial cells from differentiated NSCs

This method greatly reduces the amount of variability between NSC isolations as opposed to traditional methods like manual isolation. In addition, four of the five included conjugates can be used to isolate neurons from glia from differentiated NSCs.

  • NSCs are sorted from the heterogeneous cell culture utilizing the included antibody conjugates to five cell surface marker

TRUSTED_SUSTAINABILITY

Specifications

Reactivity Human
Quantity 10 Tests
For Use With (Application) Designed to allow the isolation of neural stem cells (NSCs) derived from Human pluripotent stem cells or the isolation of neurons and glial cells from differentiated NSCs
Content And Storage 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.