Learn More
Description
- Far-red fluorescent DNA dye
- Cell permeable
- Used to stain nucleic acids in live or fixed cells
- Has an excitation wavelength maxima of 598/646nm
Specifications
Specifications
| Color | Far-red |
| Concentration | 5 mM |
| Content And Storage | Store undiluted at 4°C |
| For Use With (Application) | Bioimaging, Immunofluorescence, Intracellular Staining (Flow Cytometry) |
| Dye Type | DNA dye |
| Excitation Wavelength Range | 598/646 nm |
| Quantity | 200 μL |
| Reactivity | Tested in Development: Human, Mouse; Reported Reactivity: Rat |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.