Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ Purified NA/LE Human BD Fc Block™

Catalog No. bdb564765 Shop All BD Biosciences Products
Click to view available options
Quantity:
0.25 mg

Catalog No. BDB564765

Supplier: BD 564765

Only null left
Add to Cart
Add to Cart

0.25mg in a 1.0mg/mL concentration

Fcγ Receptors belong to the immunoglobulin superfamily and are expressed at varying levels in multiple cell lineages including high expression in myeloid and B cells. The major functions of Fc receptors are protective functions of the immune system. There are multiple different types of Fc receptors reflecting a variety of different biological activities, which are modulated when they are aggregated by multivalent antigen-antibody complexes.

While normally serving important physiological roles in the immune system, Fc Receptors can also be the cause of nonspecific, false-positive antibody staining of cells. Human BD Fc Block™ is designed and formulated to block or significantly reduce potential non-specific antibody staining caused by receptors for IgG that may be encountered in various applications including the flow cytometric analysis of human cells.
Moreover, it can increase the specificity of antibody labeling of extremely rare target cells such as antigen-specific B cells, fetal cells in maternal blood, hematopoietic progenitor cells, or disseminated epithelial tumor cells.

Order Info

This preparation contains no preservatives, thus it should be handled under aseptic conditions.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.0 mg/mL
Quantity 0.25 mg
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001ng/μg) of protein as determined by the LAL assay.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.