Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Phospholipid-binding Protein, APC Annexin V
Shop All BD Biosciences Products
Click to view available options
Quantity:
100 Tests
200 Tests
Description
- Conjugated to APC
- Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes
Flow Cytometry

Specifications
Specifications
Quantity | 200 Tests |
Sample Volume | 5 μL |
Storage Buffer | Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide |
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Conjugate | APC (Allophycocyanin) |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction