Learn More
BD Ratiometric Calcium Assay Kit
Catalog No. p-4003688
Click to view available options
Quantity:
10 Preps
100 Preps
Includes
Ratiometric Calcium Indicator, lyophilized 10X Signal Enhancer
Involves single dye addition step with no subsequent washing
  • For homogeneous measurement of intracellular calcium changes caused by activation of G-protein coupled receptors or calcium channels
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
For Use With (Application) Kit allows homogeneous measurement of intracellular calcium changes caused by activation of G-proteins coupled receptors or calcium channels
Includes 1 vial ratiometric calcium indicator, 10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer
Quantity 10 Preps
Type Detection Assay Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.