Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD TMB Substrate Reagent Set

Catalog No. bdb555214
Click to view available options
Format:
ELISA Set

Includes

  • Substrate reagent A, 300mL
  • Substrate reagent B, 300mL

For use in BD OptEIA™ ELISA sets. Substrate Reagent A, which is clear and colorless, contains hydrogen peroxide. Substrate Reagent B is colorless to a very light amber solution containing 3,3', 5,5' tetramethylbenzidine (TMB) in an organic solvent.

  • Bring reagents to room temperature prior to use (allow 30 minutes for this process)
  • Discard any remaining working solutions after use
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Substrate
Format ELISA Set
For Use With (Application) ELISA
Kit Contents Substrate reagent A, 300mL
Substrate reagent B, 300mL
Content And Storage 2°C to 8°C

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.