Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDH2 Antibody (OTI2G1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB272067APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272067APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272067APC Supplier Novus Biologicals Supplier No. NBP272067APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BDH2 Monoclonal specifically detects BDH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen BDH2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2G1
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 3-hydroxybutyrate dehydrogenase, type 2, dehydrogenase/reductase (SDR family) member 6, Dehydrogenase/reductase SDR family member 6, DHRS6, EC 1.1.1.-, EC 1.1.1.30, EFA6R, FLJ13261, Oxidoreductase UCPA, PRO20933, R-beta-hydroxybutyrate dehydrogenase, SDR15C1,3-hydroxybutyrate dehydrogenase type 2, short chain dehydrogenase/reductase family 15C, member 1, UCPA-OR, UNQ6308
Gene Symbols BDH2
Host Species Mouse
Immunogen Full length recombinant protein of human BDH2 (NP_064524) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 56898.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.