Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDNF Antibody (35909), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB304413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304413 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304413 Supplier Novus Biologicals Supplier No. FAB2481D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BDNF Monoclonal antibody specifically detects BDNF in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen BDNF
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 35909
Conjugate DyLight 350
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF, His129-Arg247, Accession # P23560
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 627
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.