Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDNF Antibody (35928.11), Novus Biologicals™
SDP

Catalog No. NB12010505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12010505 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12010505 Supplier Novus Biologicals Supplier No. NB12010505
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen BDNF
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 35928.11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 8 - 25 ug/mL
Gene Alias abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Gene Symbols BDNF
Host Species Mouse
Immunogen Recombinant human BDNF (rhBDNF) expressed in Sf 21 insect cells.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 627
Test Specificity Monoclonal Anti-Human BDNF will neutralize the biological activity of rhBDNF. This antibody shows < 2% cross-reactivity with rhbeta-NGF, rhNT-3 and rhNT-4 based on ELISA and immunoblotting.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.