Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB50249A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50249A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50249A750 Supplier Novus Biologicals Supplier No. NB500249AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Beclin 1 Polyclonal specifically detects Beclin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen Beclin 1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Gene Symbols BECN1
Host Species Rabbit
Immunogen Internal synthetic peptide to human Beclin 1, within residues 1-100 [UniProtNo. Q14457].
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8678.0
Target Species Human, Mouse, Rat, SARS-CoV
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.