Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beclin 1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 46 publications
Supplier: Novus Biologicals NB11087318SS
Description
Beclin 1 Polyclonal specifically detects Beclin 1 in Human, Mouse, Rat, Porcine, Alligator, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Beclin 1 | |
Polyclonal | |
Unconjugated | |
PBS, 0.02 Sodium Azide with 0.02% Sodium Azide | |
ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30 | |
Rabbit | |
51 kDa | |
0.025 mL | |
Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors | |
8678 | |
Human, Mouse, Rat, Pig, Alligator, Canine, Primate | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
1 mg/mL | |
Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50 - 1:400, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50 - 1:400, KnockDown Validated reported in scientific literature (PMID 29297744) | |
Q14457 | |
BECN1 | |
Synthetic peptide made to an internal portion of human Beclin 1 (within residues 150-300). [Swiss-Prot: Q14457] | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat (96%), monkey, bovine (97%), porcine. Canine reactivity reported in scientific literature (PMID: 24027311). Possible reactivity to Chicken (87%), Xenopus laevis (79%), and Xenopus tropicalis (77%) (not yet validated). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction