Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beclin 1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 46 publications
$304.50 - $499.50
Specifications
Antigen | Beclin 1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50 - 1:400, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50 - 1:400, KnockDown Validated reported in scientific literature (PMID 29297744) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11087318T
![]() |
Novus Biologicals
NB11087318SS |
0.025 mL |
Each for $304.50
|
|
|||||
NB11087318
![]() |
Novus Biologicals
NB11087318 |
0.05 mL |
Each for $499.50
|
|
|||||
Description
Beclin 1 Polyclonal specifically detects Beclin 1 in Human, Mouse, Rat, Porcine, Alligator, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Beclin 1 | |
Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50 - 1:400, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50 - 1:400, KnockDown Validated reported in scientific literature (PMID 29297744) | |
Polyclonal | |
Rabbit | |
Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors | |
PBS, 0.02 Sodium Azide with 0.02% Sodium Azide | |
ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30 | |
BECN1 | |
IgG | |
Affinity Purified | |
Human, mouse, rat (96%), monkey, bovine (97%), porcine. Canine reactivity reported in scientific literature (PMID: 24027311). Possible reactivity to Chicken (87%), Xenopus laevis (79%), and Xenopus tropicalis (77%) (not yet validated). |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Pig, Alligator, Canine, Primate | |
Q14457 | |
8678 | |
Synthetic peptide made to an internal portion of human Beclin 1 (within residues 150-300). [Swiss-Prot: Q14457] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
51 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title