Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB357686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357686 Supplier Novus Biologicals Supplier No. NBP176648PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Beclin 1 Polyclonal antibody specifically detects Beclin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Beclin 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the center of human Beclin 1. The immunogen is located between 40-90 amino acids of Beclin 1.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8678
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.