Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 1, p Ser234 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229654 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229654 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229654 Supplier Novus Biologicals Supplier No. NBP229654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Beclin 1 Polyclonal specifically detects Beclin 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Beclin 1 (p Ser234)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:250
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. Q14457
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Gene Symbols BECN1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser234 conjugated to KLH. Accession # Q14457. Accession # Q14457
Molecular Weight of Antigen 60 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8678
Test Specificity Specific for the ∼60k beclin1 protein phosphorylated at Ser234.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.