Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11060982T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060982T 0.025 mL
NB11060982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11060982T Supplier Novus Biologicals Supplier No. NB11060982SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Beclin 2 Polyclonal specifically detects Beclin 2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Beclin 2
Applications Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200-1:500
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. A8MW95
Gene Alias BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1
Gene Symbols BECN2
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Beclin 2 protein. [Uniprot # A8MW95]
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 441925
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.