Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11060984 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060984 0.1 mL
NB11060984T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11060984 Supplier Novus Biologicals Supplier No. NB11060984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Beclin 2 Polyclonal specifically detects Beclin 2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Beclin 2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200-1:500, Simple Western 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26506105)
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. A8MW95
Gene Alias BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1
Gene Symbols BECN2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 200-300) of the human Beclin 2 protein. [UniProt# A8MW95]
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 441925
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.