Learn More
Description
Specifications
Specifications
| Antigen | Beclin 2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200-1:500, Simple Western 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26506105) |
| Formulation | PBS and 30% Glycerol with 0.1% Sodium Azide |
| Gene Accession No. | A8MW95 |
| Gene Alias | BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1 |
| Gene Symbols | BECN2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
