Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB339070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339070 Supplier Novus Biologicals Supplier No. NB11060984PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Beclin 2 Polyclonal antibody specifically detects Beclin 2 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Beclin 2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 200-300) of the human Beclin 2 protein. [UniProt# A8MW95]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 441925
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.