Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclomethasone dipropionate, 97%, Thermo Scientific Chemicals

Catalog No. p-7077865
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5534-09-8
Molecular Formula C28H37ClO7
Molecular Weight (g/mol) 521.04
InChI Key KUVIULQEHSCUHY-XYWKZLDCSA-N
Synonym beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm
PubChem CID 134129500
IUPAC Name [2-[(8R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMILES CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)Cl)O)C)C)OC(=O)CC

Specifications

Melting Point 208.0°C
Loss on Drying 1% max. (105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 250 mg
Formula Weight 521.04
Percent Purity 97%
Chemical Name or Material Beclomethasone dipropionate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 226-886-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.