Learn More
Bel-Art™ Heavy Duty Pail
Polyethylene pails have wide rims that hold their shape under heavy loads
$353.51
Specifications
Product Type Heavy Duty Pail
Graduations Liters
Material Polyethylene
Color White
View More Specs
Products 2
Catalog Number Mfr. No. dimCapacityMetric HeightMetric Diameter (Metric)Top Price Quantity & Availability  
Catalog Number Mfr. No. dimCapacityMetric HeightMetric Diameter (Metric)Top Price Quantity & Availability  
22-163659
View Documents
Bel-Art™
H168060000
Each for $353.51
Only null left
Add to Cart
22-020073
View Documents
Bel-Art™
H168050000
24.4 cm N/A N/A N/A
Description
Description
  • Features include a heavy-gauge, plastic-coated metal bail and a pronounced spout
  • Recessed hand grip on the bottom aids in safe handling and pouring
  • Graduations are in liters and molded on the interior
Specifications
Specifications
Heavy Duty Pail
Liters
Polyethylene
White
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.