Learn More
Bel-Art™ Large Pails
Catalog No. p-3273220 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Capacity (Metric):
7.6 L
13.2 L
Material:
Polypropylene
This item is not returnable. View return policy
This item is not returnable. View return policy
Rugged pails with fitted lids/covers and steel-plated bail handles
  • Translucent white pails allow for easier viewing of fluid level
  • Polypropylene is steam autoclavable (121°C), Polyethylene is not autoclavable
  • 8-qt./7.6L pail has removable steel-plated support ring and bail handle
  • 14-qt./13.2L pail has steel-plated swinging bail handle with grip, no support ring
  • Large pails meets US Specification MIL-P36257A
Specifications
Product Type Pail
Autoclavable Not Autoclavable
Certifications/Compliance US, MIL-P36257A, FDA
Color Yellow
Description 2.99 gal. Pail
Handle Material Plated Steel
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.