Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Transpette™ Eight Channel Transfer Pipettor

Catalog No. p-313740 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Material:
Polyethylene
Polypropylene
Packaging Type:
Bulk
Individually Wrapped
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Material Packaging Type
13-682-105 Sterile Polyethylene Individually Wrapped
13-682-104 Non-sterile Polypropylene Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-682-105 Supplier Bel-Art™ Supplier No. F378730001
Only null left
Add to Cart
Add to Cart

For washing plates, feeding, and supernatant aspiration from microwell plates

  • 8-Channel with 600μL capacity per channel
  • Made of translucent plastic
  • Discard after hot radioactive uses to avoid radiation contamination of expensive liquid handling devices
  • Non-autoclavable
  • Available with sterile and non-sterile options

Specifications

Material Polyethylene
Sterility Sterile
For Use With (Application) For washing plates, feeding, and supernatant aspiration from microwell plates
Quantity 100/Cs.
Packaging Type Individually Wrapped
Color Natural
Volume (Metric) 600 μL
Length (Metric) 180 mm
Type Transfer Pipet
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.