Learn More
Wilmad-LabGlass™ SP Scienceware™ 5mm O.D. Thin Walled Economy NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
Designed for routine use in all NMR spectrometers, Wilmad Economy NMR Tubes are guaranteed to fit tightly in any spinner turbine and have zero NMR background
$19.00 - $104.00
Specifications
Thickness (Metric) Wall 0.43 mm
Diameter (Metric) Outer 4.947±0.019 mm
View More Specs
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Products 14
Catalog Number Mfr. No. Model Length (Metric) Field Strength Price Quantity & Availability  
Catalog Number Mfr. No. Model Length (Metric) Field Strength Price Quantity & Availability  
16-800-273
View Documents
Wilmad-LabGlass™
661009575
WG-1206-7-5 178 mm 100MHz
Pack of 5 for $19.00
Add to cart
16-800-274
View Documents
Wilmad-LabGlass™
661009585
WG-1206-8-5 203.2 mm 100MHz
Pack of 5 for $19.00
Add to cart
16-800-318
View Documents
Wilmad-LabGlass™
663009575
WG-1226-7-5 178 mm 300MHz
Pack of 5 for $24.00
Add to cart
16-800-319
View Documents
Wilmad-LabGlass™
663009585
WG-1226-8-5 203.2 mm 300MHz
Pack of 5 for $32.50
Add to cart
16-800-405 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Wilmad-LabGlass™
664009575
WG-1228-7-5 178 mm 400MHz
Pack of 5 for $27.75
Add to cart
16-800-406
View Documents
Wilmad-LabGlass™
664009585
WG-1228-8-5 203.2 mm 400MHz
Pack of 5 for $33.00
Add to cart
16-800-443 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Wilmad-LabGlass™
665009575
WG-1235-7-5 178 mm 500MHz
Pack of 5 for $39.50
Add to cart
16-800-444
View Documents
Wilmad-LabGlass™
665009585
WG-1235-8-5 203.2 mm 500MHz
Pack of 5 for $49.00
Add to cart
16-800-471 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Wilmad-LabGlass™
666009575
WG-1241-7-5 178 mm 600MHz
Pack of 5 for $43.50
Add to cart
16-800-472
View Documents
Wilmad-LabGlass™
666009585
WG-1241-8-5 203.2 mm 600MHz
Pack of 5 for $55.00
Add to cart
16-800-489
View Documents
Wilmad-LabGlass™
667009575
WG-1242-7-5 178 mm 700MHz
Pack of 5 for $91.00
Add to cart
16-800-490
View Documents
Wilmad-LabGlass™
667009585
WG-1242-8-5 203.2 mm 700MHz
Pack of 5 for $104.00
Add to cart
16-800-290
View Documents
Wilmad-LabGlass™
662009575
WG-1208-7-5 178 mm 200MHz
Pack of 5 for $19.00
Add to cart
16-800-291
View Documents
Wilmad-LabGlass™
662009585
WG-1208-8-5 203.2 mm 200MHz
Pack of 5 for $21.25
Add to cart
Description
Description
Manufactured in an ISO 9001:2008 USA facility
  • Best O.D. tolerance in the industry and a 30% thicker wall than any competitors' product
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
  • Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A
  • Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes
Specifications
Specifications
0.43 mm
4.947±0.019 mm