Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heathrow Scientific™ Benchtop Dispensing Bin

Click to view available options
Style:
Dual Compartments
Single Compartment
Description
- Lid remains open when tilted back
- Non-skid rubber feet
- Pre-drilled holes for wall mounting
- Mounting screws included for the option of fixing to a wall or raised surface
- Dual compartment bin includes a removable divider
Specifications
Specifications
Type | Dispensing Bin |
Material | Acrylic |
Color | Neon Blue |
Autoclavable | Not Autoclavable |
Closure Type | Lid |
Description | Benchtop Dispensing Bin |
For Use With (Application) | Storage and Organization of Lab Supplies, Accessories |
Holds | Lab supplies, accessories |
Style | Single Compartment |
Quantity | 1 Ea. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction