Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Benchtop Meters: pHBasic Series

Catalog No. p-7082534 Shop All Sartorius Products
Change view
Click to view available options
Type:
pHBasic Meter Only
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-558-062 pHBasic Meter Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-558-062 Supplier Sartorius Supplier No. PHBASICA

For basic everyday pH routines

  • Simple-to-use with classic interface design and intuitive icons
  • Classic no-fuss menus and intuitive operation
  • Works faster due to optimized algorithms with harmonized measuring system of sensor with meter
  • Large LCD displays with simultaneous pH/mV and temperature
  • Reliable with understandable status icons and real time indicators like ElectCheck, and Stability Control
  • Automatic temperature compensation
  • Automatic buffer recognition- up to 16 buffer sets
  • Easy 1-2-3 point Calibration
  • German quality
  • Quick access to electrode calibration data
  • At a glance know if sensor efficiency and calibration integrity are intact

Note: ElectCheck is a sensor evaluation and calibration reminder. The sensor evaluation shows the electrode quality in increments. The integrity criteria are slope, zero point and reaction time. When the time interval has elapsed, an optical and an acoustical signal is activated.

TRUSTED_SUSTAINABILITY

Specifications

Description Meter, Power Supply, Cover, Manual (3pt Cal and Electcheck)
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.3, ±1
Accuracy (pH) ±0.1, ±0.01, ±0.005
Includes pHBasic Meter, Power Supply, BNC connector, Transparent cover, Manual, 4 AA batteries
For Use With (Application) Sample and Buffer Preparation - R&D environments
Range (pH) -2.0 to 20.0, -2.00 to 20.00, -2.000 to 19.999
Range (mV) ±1200.0, ±2000
Calibration Points 3
Temperature Range (Metric) -5°C to +105°C
Display Type LCD, Large with Simultaneous pH/mV and Temperature
Warranty 2 Years
Type pHBasic Meter Only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.