Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6-chloronicotinate, 97%, Thermo Scientific™

Catalog No. AC454640250
Click to view available options
Quantity:
5 g
25 g
100 g

Chemical Identifiers

CAS 49608-01-7
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD00082739
InChI Key ILDJJTQWIZLGPO-UHFFFAOYSA-N
PubChem CID 2799611
IUPAC Name ethyl 6-chloropyridine-3-carboxylate
SMILES CCOC(=O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 26.0°C to 30.0°C
Boiling Point 98.0°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Synonym ethyl 6-chloronicotinate, ethyl 6-chloro-3-pyridinecarboxylate, ethyl6-chloronicotinate, 6-chloro-nicotinic acid ethyl ester, 6-chloronicotinic acid ethyl ester, ethyl 6-chloro nicotinate, ethyl 2-chloro-5-pyridinecarboxylate, 3-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem1415, pubchem15179
Formula Weight 185.61
Percent Purity 97%
Chemical Name or Material Ethyl 6-chloronicotinate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.