Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Benzamil
GREENER_CHOICE

Catalog No. 338010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-801-0 10 mg
33-805-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-801-0 Supplier Tocris Bioscience™ Supplier No. 3380/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

NCX inhibitor, Deg/ENaC channel blocker; amiloride (Cat. No. 0890) derivative

Na+/Ca2+ exhanger (NCX) inhibitor (IC50 ∽ 100 nM); TRPP3 channel blocker (IC50 = 1.1 μM for inhibition of Ca2+-activated TRPP3 channel activity). Also non-selective Deg/ENaC family blocker; reduces mechanosensitivity of colonic afferents. More potent derivative of amiloride (Cat. No. 0890).

Chemical Identifiers

CAS 161804-20-2
Molecular Formula C13H14ClN7O·HCl

Specifications

Quantity 10 mg
Formula Weight 356.21
Purity >98%
Chemical Name or Material Benzamil
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.