Learn More
Benzenesulfinic acid sodium salt, 98%
Description
It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidants. Store in cool, dry place in a tightly sealed container.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 25 g |
| Beilstein | 3598405 |
| Solubility Information | Soluble in water. |
| Formula Weight | 164.16 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzenesulfinic acid sodium salt |
Safety and Handling
| EINECSNumber | 212-842-8 |
| RTECSNumber | DA9135000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.