Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzidines Mixture (For EPA 605, 625)

Catalog No. usus105n
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-US-105N 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-US-105N Supplier Agilent Technologies Supplier No. US105N1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Page 304C, Designated A2LA-Certified product.

Chemical Identifiers

CAS 92-87-5
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
InChI Key HFACYLZERDEVSX-UHFFFAOYSA-N
PubChem CID 7111
ChEBI CHEBI:80495
IUPAC Name 4-(4-aminophenyl)aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)N)N

Specifications

Packaging Ampule
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methanol:Benzidine3, 3«-Dichlorobenzidine
Chemical Name or Material Benzidines Mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.