Learn More
Benzophenone hydrazone, 98+%
Description
An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.
Solubility
Soluble in ether, benzene, chloroform and other organic solvents.
Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Keep refrigerated. (Store below 4 C/39 F.)
Specifications
Specifications
| Melting Point | 96°C to 100°C |
| Density | 1.1 |
| Boiling Point | 225°C to 230°C (55 mmHg) |
| Flash Point | 146°C (294°F) |
| Odor | Characteristic |
| Quantity | 50 g |
| UN Number | UN2811 |
| Beilstein | 1910177 |
| Solubility Information | Soluble in ether,benzene,chloroform and other organic solvents. |
| Formula Weight | 196.25 |
| Show More |
Safety and Handling
| Hazard Category | H301-H317 |
| Hazard Statement | GHS H Statement H301-H317 Toxic if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 226-321-8 |
| RTECSNumber | PC4954487 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.