Alfa Aesar™ Benzylamine, 98+%

Manufacturer: Alfa Aesar™ A1099730

Quantity 250g

 View more versions of this product

Catalog No. AAA1099730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-46-9
    Molecular Formula C7H9N
    Formula Weight 107.16
    MDL Number MFCD00008106
    UN Number UN2735
    Beilstein 741984
    Merck Index 14,1125
    Chemical Name or Material Benzylamine

    Specifications

    Formula Weight 107.16
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 100-46-9
    Molecular Formula C7H9N
    Merck Index 14,1125
    Health Hazard 2 GHS H Statement
    H314-H302-H312-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.
    Melting Point −43°C
    MDL Number MFCD00008106
    UN Number UN2735
    Beilstein 741984
    Health Hazard 1 Danger
    Boiling Point 184° to 185°C
    Quantity 250g
    Flash Point 72°C (162°F)
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    RTECS Number DP1488500
    Chemical Name or Material Benzylamine
    Assay ≥98%

    Description

    Applications
    Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.

    Solubility
    Miscible with water, ethanol, diethyl ether, acetone, benzene and chloroform.

    Notes
    Air sensitive. Incompatible with strong oxidizing agents and strong acids.

    GHS H Statement
    H314-H302-H312-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit