Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylamine hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
Plastic bottle
3287-99-8
C7H9N·ClH
143.62
MFCD00012852
XKXHCNPAFAXVRZ-UHFFFAOYSA-N
benzylamine hydrochloride,phenylmethanamine hydrochloride,benzenemethanamine, hydrochloride,benzylammonium chloride,benzylamine, hydrochloride,usaf el-82,benzenemethanamine, hydrochloride 1:1,benzylamine hcl,benzylaminehydrochloride,benzyl amine hcl salt
2724127
phenylmethanamine;hydrochloride
C1=CC=C(C=C1)CN.Cl
3287-99-8
C7H9N·ClH
143.62
MFCD00012852
XKXHCNPAFAXVRZ-UHFFFAOYSA-N
benzylamine hydrochloride,phenylmethanamine hydrochloride,benzenemethanamine, hydrochloride,benzylammonium chloride,benzylamine, hydrochloride,usaf el-82,benzenemethanamine, hydrochloride 1:1,benzylamine hcl,benzylaminehydrochloride,benzyl amine hcl salt
2724127
phenylmethanamine;hydrochloride
C1=CC=C(C=C1)CN.Cl
Color White
Melting Point 259°C to 264°C
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CH2NH2·HCl
Beilstein 12, 1017
Merck Index 14, 1125
Solubility Information Solubility in water: soluble
Formula Weight 143.62
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material Benzylamine hydrochloride, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-943-6

RTECSNumber : DP5425000

TSCA : TSCA