Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltri-n-butylammonium bromide, 99%

Catalog No. AAB2394118
Click to view available options
Quantity:
25 g
50 g
250 g

CAS: 25316-59-0 | C19H34BrN | 356.392 g/mol

Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-59-0
Molecular Formula C19H34BrN
Molecular Weight (g/mol) 356.392
MDL Number MFCD00011848
InChI Key UDYGXWPMSJPFDG-UHFFFAOYSA-M
Synonym benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
PubChem CID 2724282
IUPAC Name benzyl(tributyl)azanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]

Specifications

Melting Point ∼175°C (decomposition)
Quantity 50 g
Beilstein 3776294
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 356.4
Percent Purity 99%
Chemical Name or Material Benzyltri-n-butylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-819-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.