Learn More
Siemens Healthineers Berichrom Antithrombin III (A) Reagent
Determination of the functional activity of antithrombin III (AT III) in human plasma.
$583.00 - $1636.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-044-707
View Documents
Siemens Healthineers
10446673
150 Tests/Kit
Pack of 6 for $583.00
Only null left
Add to Cart
23-044-706
View Documents
Siemens Healthineers
10446672
500 Tests/Kit
Pack of 6 for $1,636.00
Only null left
Add to Cart
Description
Description
Berichrom Antithrombin III (A) is a chromogenic activity assay for the detection of hereditary or acquired antithrombin deficiency, thrombophilia, and the monitoring of patients undergoing substitution therapy. The heparin cofactor-independent lyophilized reagent uses bovine thrombin substrate and exhibits no interference with anti-FXa anticoagulants, e.g., rivaroxaban.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.