Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Berichrom Protein C Reagent
Quantative determination of protein C activity as an aid in the diagnosis of inherited and acquired deficiencies.
$891.00 - $1262.00
Specifications
Type | Hemostasis Reagent |
---|---|
For Use With (Application) | Thrombophilia Testing |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-044-696
|
Siemens Healthineers
10446499 |
3 x 10 mL |
Pack of 3 for $1,262.00
|
|
|||||
23-044-697
|
Siemens Healthineers
10446500 |
4 x 5 mL |
Pack of 4 for $891.00
|
|
|||||
Description
Berichrom Protein C, a chromogenic functional activity assay, is used for the detection of hereditary or acquired protein C deficiencies in conjunction with other methods (antigenic determination, protein C clotting method) for the differential diagnosis of different protein C deficiency states. The assay is also used for the monitoring of substitution therapy with protein C concentrates in congenital protein C deficiency. The Berichrom Protein C assay is less susceptible to interfering substances than a clotting assay.Specifications
Hemostasis Reagent | |
Thrombophilia Testing |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title