SHOWA™ Fuzzy Duck™ Gloves

Cool and abrasion resistant

Manufacturer: SHOWA™ 96210

 View more versions of this product

Catalog No. 11-388-131C

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Exterior Finish Textured
    Glove Cuff Style Slip-on
    For Use With (Application) small parts handling, general maintenance, shipping and assembly
    Material PVC
    Liner Material Cotton Jersey
    Size 10
    Coated Area Full
    Color Yellow
    Cut Resistance Rating Level 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit