Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Nitri-Dex™ Flock-Lined Nitrile Gloves

Catalog No. 191501293 Shop All Showa Best Glove Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy

Catalog No. 19-150-1293

Supplier: SHOWA™ 707FL09

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unsupported, flock-lined nitrile coated gloves feature a lasting grip and provide protection from oils, hydrocarbons, grease and abrasion.

  • Ideal for jobs requiring comfort, tactility, and contact with chemical hazards
  • Flocked cotton liner ensures easy donning and doffing
  • Rolled cuff prevents debris from entering
  • Lightweight nitrile material helps reduce hand fatigue

Specifications

ANSI Abrasion Rating ANSI 1
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) CE
Chlorinated No
Color Blue
Latex Free Yes
Material Nitrile
Size Large
Thickness (English) 9 mil
Thickness (Metric) 0.23 mm
EN388 Standard EN 388:2003 1001
EN 421 No
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Manufacturing, laboratories, paint applications, fuel handling, food processing, janitorial, agricultural chemicals, refineries, light duty maintenance and clean up, paint applications, poultry processing
Glove Cuff Style Gauntlet
Grip Design Tractor tread
Liner Material Cotton Flock
Lining Lined
Model 707FL-09
Series SHOWA™ 707FL
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.