Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ Zorb-IT Ultra Work Gloves
Shop All Showa Best Glove Products
Click to view available options
Size:
7
8
9
10
11
Specifications
Specifications
| Color | Gray, Yellow |
| Cut Resistance Rating | ANSI A3 |
| Material | Aramid Fiber, Lycra |
| Product Type | Cut-resistant Gloves |
| Size | 7 |
| For Use With (Application) | Automotive, construction, glass, metallurgy |
| Glove Cuff Style | Elasticated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction