Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Heavy Duty PVC Coated Seamless Knit Cotton Gloves

Catalog No. p-144818 Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

PVC-coating minimizes the absorption of water, oil, and grease with a rough-texture to enhance grip capabilities.

  • PVC-coating ensures breathability
  • Easy donning and doffing with slip-on cuff
  • Minimizes absorption of water, oil, and grease
  • Textured to enhance grip capabilities

Specifications

Coated Area Full Glove
Construction Seamless Knit
Coating Material PVC
Color Yellow
Exterior Finish Fully Textured
Hand Specific
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Chemical Spray/Treatment, Utility Work, Intricate Parts Handling, and Oil-Coated Pieces
Glove Cuff Style Continuous Knit
Liner Material Cotton
Packaging Type Polybag
Series 960
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.