Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Strapper™ Gloves

Catalog No. 19014538 Shop All Showa Best Glove Products
Click to view available options
Size:
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Excellent replacement for leather driver's style glove

Nitrile laminated in palm area for maximum cut, puncture, and snag resistance combined with flexibility.

  • Gunn-cut wing-thumb construction with shirred back gives extra comfort.
  • Sanitized™ liners retard deteriorating effects of perspiration, mold and mildew
  • Gloves have breathable, striped cotton back and are machine washable

Specifications

Back Material Cotton
Coated Area Palm, Index Finger
Coating Material Nitrile
Color Black, Natural
Exterior Finish No Texture
Hand Specific
Product Type Dip-coated Gloves
Size 9
For Use With (Application) Assembly, Inspection, Fabrication, and Light/Medium Material Handling
Glove Cuff Style Open
Series 2790
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.