Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Super Flex™ Gloves

Catalog No. p-3139304 Shop All Showa Best Glove Products
Click to view available options
Glove Cuff Style:
Gauntlet
This item is not returnable. View return policy
This item is not returnable. View return policy

Low-temperature-resistant PVC formulation soft and supple even in cold conditions

Provide excellent abrasion resistance and protection from a broad range of chemicals.

  • Cotton jersey liner enhances worker comfort and glove flexibility
  • Yellow color provides a high-contrast, visual warning of possible contamination
  • Fully coated
  • Smooth finish
  • Sanitized™

Specifications

Coated Area Full
Color Yellow
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Material PVC
Size 10
Sterility Sterile
For Use With (Application) manufacturing, light duty maintenance, refining, Petrochemicals, mining, construction, utilities
Glove Cuff Style Gauntlet
Liner Material Cotton Jersey
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.