Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bestrophin 1 Antibody (E6-6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300164 0.1 mL
NB300164SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300164 Supplier Novus Biologicals Supplier No. NB300164
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 42 publications

Bestrophin 1 Monoclonal specifically detects Bestrophin 1 in Canine, Human, Mouse, Pig, Primate, Rat, Rat (Negative) samples. It is validated for Dual RNAScope ISH-IHC, Gene Knock-OutC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot.

Specifications

Antigen Bestrophin 1
Applications Dual RNAScope ISH-IHC, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot
Classification Monoclonal
Clone E6-6
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry reported in scientific literature (PMID 30048622), Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24345323), Immunohistochemistry-Frozen, Proximity Ligation Assay reported in scientific literature (PMID 27519691), Knockout Validated, Dual RNAscope ISH-IHC
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O76090
Gene Alias ARB, BEST, Best disease, bestrophin 1, bestrophin-1, BMD, TU15B, vitelliform macular dystrophy 2, Vitelliform macular dystrophy protein 2, VMD2RP50
Gene Symbols BEST1
Host Species Mouse
Immunogen Synthetic peptide conjugated to KLH corresponding to the C-terminus of human Bestrophin 1 (KDHMDPYWALENRDEAHS) [Uniprot: O76090]
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 7439
Test Specificity Bestrophin (E6-6)
Target Species Canine, Human, Mouse, Pig, Primate, Rat, Rat (Negative)
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.