Learn More
Description
Specifications
Specifications
| Antigen | Bestrophin 1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA |
| Classification | Monoclonal |
| Clone | E6-6 |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry reported in scientific literature (PMID 30048622), Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24345323), Immunohistochemistry-Frozen, Proximity Ligation Assay reported in scientific literature (PMID 27519691), Knockout Validated, Dual RNAscope ISH-IHC |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O76090 |
| Gene Alias | ARB, BEST, Best disease, bestrophin 1, bestrophin-1, BMD, TU15B, vitelliform macular dystrophy 2, Vitelliform macular dystrophy protein 2, VMD2RP50 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
